Merck
  • Mutations in ASPH cause facial dysmorphism, lens dislocation, anterior-segment abnormalities, and spontaneous filtering blebs, or Traboulsi syndrome.
Mutations in ASPH cause facial dysmorphism, lens dislocation, anterior-segment abnormalities, and spontaneous filtering blebs, or Traboulsi syndrome.
American journal of human genetics (2014-04-29)
Nisha Patel, Arif O Khan, Ahmad Mansour, Jawahir Y Mohamed, Abdullah Al-Assiri, Randa Haddad, Xiaofei Jia, Yong Xiong, André Mégarbané, Elias I Traboulsi, Fowzan S Alkuraya
ABSTRAKT
We have previously described a syndrome characterized by facial dysmorphism, lens dislocation, anterior-segment abnormalities, and spontaneous filtering blebs (FDLAB, or Traboulsi syndrome). In view of the consanguineous nature of the affected families and the likely autosomal-recessive inheritance pattern of this syndrome, we undertook autozygosity mapping and whole-exome sequencing to identify ASPH as the disease locus, in which we identified two homozygous mutations. ASPH encodes aspartyl/asparaginyl β-hydroxylase (ASPH), which has been found to hydroxylate aspartic acid and asparagine residues on epidermal growth factor (EGF)-domain-containing proteins. The truncating and missense mutations we identified are predicted to severely impair the enzymatic function of ASPH, which suggests a possible link to other forms of ectopia lentis given that many of the genes implicated in this phenotype encode proteins that harbor EGF domains. Developmental analysis of Asph revealed an expression pattern consistent with the proposed link to the human syndrome. Indeed, Asph-knockout mice had a foreshortened snout, which corresponds to the facial abnormalities in individuals with Traboulsi syndrome. These data support a genetic basis for a syndromic form of ectopia lentis and the role of aspartyl hydroxylation in human development.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
Hypromellose, meets USP testing specifications
USP
Hypromeloza, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture