Merck
  • B-lymphoblastic leukemia/lymphoma: overexpression of nuclear DNA repair protein PARP-1 correlates with antiapoptotic protein Bcl-2 and complex chromosomal abnormalities.
B-lymphoblastic leukemia/lymphoma: overexpression of nuclear DNA repair protein PARP-1 correlates with antiapoptotic protein Bcl-2 and complex chromosomal abnormalities.
Human pathology (2014-05-27)
Payam Pournazari, Ruth F Padmore, Farid Kosari, Peter Scalia, Meer-Taher Shahbani-Rad, Sami Shariff, Douglas J Demetrick, Mark Bosch, Adnan Mansoor
ABSTRAKT
Poly(ADP-ribose) polymerase-1 (PARP-1) and Bcl-2 are emerging as therapeutic targets in various cancers. The former is a DNA repair protein associated with genomic stability and apoptosis, whereas the latter is an antiapoptotic protein having a DNA repair function through inhibition of PARP-1. Because genomic stability is critical for prognosis in B-lymphoblastic leukemia/lymphoma (B-ALL), we studied the expression of PARP-1 and Bcl-2 proteins in patients with B-ALL of different ages and compared the results with cytogenetic data. The PARP-1 protein was overexpressed in about two-thirds (61%) of patients with B-ALL. It had a nuclear location, whereas Bcl-2 protein was cytosolic. Expression of the 2 proteins showed a highly positive correlation (ρ = 0.367; P < .001). Overexpression of PARP-1 correlated with a complex karyotype (P = .030), and this correlation remained significant for coexpression of PARP-1 and Bcl-2 proteins (χ(2) = 7.498; P = .024) as well as after exclusion of pediatric patients (n = 9, P = .042). Overexpression of PARP-1 was not significantly more common in diploid versus aneuploid karyotypes (50% versus 59%, P = .610). The PARP-1 protein showed no correlation with specific chromosomal abnormalities associated with prognosis in B-ALL, as defined by the World Health Organization. In conclusion, high expression of the PARP-1 protein among patients with B-ALL is related to a complex karyotype and Bcl-2 positivity. Although these findings require validation in a larger population, the observations will be valuable in planning therapeutic trials (such as of PARP inhibitors and BH3 mimetics).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
o-Xylene, analytical standard
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder