Merck
Involvement of cAMP/EPAC/TRPM2 activation in glucose- and incretin-induced insulin secretion.
Diabetes (2014-05-16)
Masashi Yosida, Katsuya Dezaki, Kunitoshi Uchida, Shiho Kodera, Nien V Lam, Kiyonori Ito, Rauza S Rita, Hodaka Yamada, Kenju Shimomura, San-e Ishikawa, Hitoshi Sugawara, Masanobu Kawakami, Makoto Tominaga, Toshihiko Yada, Masafumi Kakei
ABSTRAKT
In pancreatic β-cells, closure of the ATP-sensitive K(+) (K(ATP)) channel is an initial process triggering glucose-stimulated insulin secretion. In addition, constitutive opening of background nonselective cation channels (NSCCs) is essentially required to effectively evoke depolarization as a consequence of K(ATP) channel closure. Thus, it is hypothesized that further opening of NSCC facilitates membrane excitability. We identified a class of NSCC that was activated by exendin (ex)-4, GLP-1, and its analog liraglutide at picomolar levels. This NSCC was also activated by increasing the glucose concentration. NSCC activation by glucose and GLP-1 was a consequence of the activated cAMP/EPAC-mediated pathway and was attenuated in TRPM2-deficient mice. The NSCC was not activated by protein kinase A (PKA) activators and was activated by ex-4 in the presence of PKA inhibitors. These results suggest that glucose- and incretin-activated NSCC (TRPM2) works in concert with closure of the KATP channel to effectively induce membrane depolarization to initiate insulin secretion. The current study reveals a new mechanism for regulating electrical excitability in β-cells and for mediating the action of glucose and incretin to evoke insulin secretion, thereby providing an innovative target for the treatment of type 2 diabetes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Acetylocysteina, United States Pharmacopeia (USP) Reference Standard
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder