Merck
Camellia sinensis (green tea) extract attenuate acrylamide induced testicular damage in albino rats.
Environmental toxicology (2013-02-01)
Heba A Yassa, Safaa M George, Abeer El Refaiy M Refaiy, Effat M Abdel Moneim
ABSTRAKT
Acrylamide is a proved toxin for testicular function, found in food when heated for long period of time. Green tea (Camellia sinensis) is a potent antioxidant; the aim of this study was to investigate the protective effect of green tea extract against the toxic effects of acrylamide in rat testes. acrylamide was administered orally to rats in different doses and also the extract of green tea was administered orally to different groups of animals in combination with the acrylamide. The weight of animals, testosterone hormone level and histopathological effect upon testicles were evaluated. Testosterone hormone level in serum, and histopathological findings were significantly improved with the co-administration of green tea extract with the acrylamide. Green tea extract reversed all the toxic effects of acrylamide even in high dose for long period (90 days). Green tea extract is a potent antioxidant antidote for the acrylamide toxic effects upon testicular function.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Supelco
Acrylamide, analytical standard
Supelco
Testosterone, VETRANAL®, analytical standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Testosterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Supelco
Acrylamide solution, 40% in H2O