Merck
SAHA inhibits the transcription initiation of HPV18 E6/E7 genes in HeLa cervical cancer cells.
Gene (2014-10-11)
Hongpeng He, Xuena Liu, Dandan Wang, Yijie Wang, Lei Liu, Hao Zhou, Xuegang Luo, Nan Wang, Bingyan Ji, Yan Luo, Tongcun Zhang
ABSTRAKT
High risk human papillomavirus (HPV) is a well recognized causative agent of cervical cancer. Suberoylanilide hydroxamic acid (SAHA) is a potential anti-cervical cancer drug; however, its effect on the expression of HPV E6 and E7 genes remains unclear. Here, we show that, in SAHA treated HeLa cells, HPV18 E6 and E7 mRNA and protein levels were reduced, HPV18 promoter activity was decreased, and the association of RNP II with HPV18 promoter was diminished, suggesting that SAHA inhibited the transcription initiation of HPV18 E6 and E7 genes. In SAHA-treated HeLa, although the level of lysine 9-acetylated histone H3 in the whole cell extracts increased obviously, its enrichment on HPV18 promoter was significantly reduced which is correlated with the down-regulation of HPV E6 and E7.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
SAHA, ≥98% (HPLC)