Merck
sec-Butyl-propylacetamide (SPD) and two of its stereoisomers rapidly terminate paraoxon-induced status epilepticus in rats.
Epilepsia (2014-11-08)
Guy Bar-Klein, Evyatar Swissa, Lyn Kamintsky, Tawfeeq Shekh-Ahmad, Rotem Saar-Ashkenazy, Yechiel Hubary, Shai Shrot, Liran Stetlander, Arik Eisenkraft, Alon Friedman, Meir Bialer
ABSTRAKT
Organophosphates (OPs) are commonly used insecticides for agriculture and domestic purposes, but may also serve as nerve agents. Exposure to OPs result in overstimulation of the cholinergic system and lead to status epilepticus (SE), a life-threatening condition that is often resistant to treatment. SE is associated with significant neuronal damage, neurocognitive dysfunction, and the development of lifelong epilepsy. Therefore, rapid termination of SE and prevention of brain damage is of high interest. Here we tested the efficacy of sec-butyl-propylacetamide (SPD) and two of its individual stereoisomers, (2S,3S)-SPD and (2R,3R)-SPD, in discontinuing OP-induced seizures. SPD is a one carbon homolog of valnoctamide, a central nervous system (CNS)-active constitutional isomer of valproic acid (VPA) corresponding amide valpromide. Rats were implanted with epidural telemetric electrodes to allow electrocorticography (ECoG) recording 24 h prior, during and 24 h after poisoning with the OP paraoxon (at a dose equivalent to 1.4 LD50 Median lethal dose). All rats were provided with antidotal treatment of atropine and toxogonin. Epileptic activity was measured using a novel automated system to evaluate the different effects of midazolam, SPD, and its individual stereoisomers in comparison to nontreated controls. Treatment with SPD or its individual stereoisomer (2S,3S)-SPD significantly shorten paraoxon-induced SE and reduced the duration of recorded pathologic activity after SE was terminated. (2S,3S)-SPD was superior to racemic-SPD in diminishing delayed pathologic epileptiform activity within the first 8 h after SE. These results suggest SPD as an efficient drug for the rapid termination of SE and pathological epileptiform activity following OP poisoning, a strategy to reduce neuronal dysfunction and the risk for lifelong epilepsy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Alkohol odwodniony, United States Pharmacopeia (USP) Reference Standard
Atropine, European Pharmacopoeia (EP) Reference Standard
USP
Glikol propylenowy, United States Pharmacopeia (USP) Reference Standard
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
2-Propylpentanoic acid
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Kwas walproinowy, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Obidoxime chloride, ≥95.0% (HPLC)
Supelco
Obidoxime chloride, analytical standard
Valproic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Atropine for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Paraoxon-ethyl, PESTANAL®, analytical standard
Sigma-Aldrich
Paraoxon-ethyl, ≥90%, oil
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%