Merck
The retinoblastoma (Rb) protein regulates ferroptosis induced by sorafenib in human hepatocellular carcinoma cells.
Cancer letters (2014-12-03)
Christophe Louandre, Ingrid Marcq, Hicham Bouhlal, Emma Lachaier, Corinne Godin, Zuzana Saidak, Catherine François, Denis Chatelain, Véronique Debuysscher, Jean-Claude Barbare, Bruno Chauffert, Antoine Galmiche
ABSTRAKT
Sorafenib is the treatment of reference for advanced hepatocellular carcinoma (HCC), the most frequent form of primary liver tumour. The loss of function of the retinoblastoma (Rb) protein is an important event during liver carcinogenesis, but it is unclear whether the Rb status modulates the response of HCC cells to sorafenib. Here, we examined this question in HCC cells with reduced levels of Rb achieved through stable RNA interference. We show that HCC cells with reduced levels of Rb exhibit a two- to threefold increase in cell death induction upon exposure to sorafenib compared with controls. Sorafenib treatment of Balb/c nude mice that received tumour xenografts derived from HCC cells with reduced Rb levels resulted in complete tumour regression in 50% of the animals treated, compared with tumour stabilization in mice that received control cells. We show that, upon exposure to sorafenib, the Rb-negative status of HCC cells promotes the occurrence of ferroptosis, a form of oxidative necrosis. The findings highlight the role of Rb in the response of HCC cells to sorafenib and the regulation of ferroptosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Nicotinamide (Niacinamide), analytical standard
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Niacynamid, United States Pharmacopeia (USP) Reference Standard