Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cytosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Cytosine, ≥99%
Adenine, European Pharmacopoeia (EP) Reference Standard
Supelco
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Ribonucleic acid from torula yeast, Type VI
Sigma-Aldrich
Ribonucleic acid diethylaminoethanol salt, Type IX
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Ribonucleic acid from baker′s yeast (S. cerevisiae)
Sigma-Aldrich
Ribonucleic acid from torula yeast, core, Type II-C
Sigma-Aldrich
Uracil, ≥99.0%
Ribonucleic acid, European Pharmacopoeia (EP) Reference Standard
Gemcitabine impurity A, European Pharmacopoeia (EP) Reference Standard
Fluorouracil impurity C, European Pharmacopoeia (EP) Reference Standard