Merck
Chlorhexidine in cosmetic products - a market survey.
Contact dermatitis (2014-09-12)
Morten S Opstrup, Jeanne D Johansen, Rossana Bossi, Michael D Lundov, Lene H Garvey
ABSTRAKT
Chlorhexidine may cause type I and type IV allergy. Some chlorhexidine-allergic individuals have been exposed in the healthcare setting as patients or healthcare workers, but for others the source of sensitization is unknown. Chlorhexidine may be used as a preservative or an antimicrobial agent in cosmetic products at a concentration up to 0.3%, as set by the European Cosmetics Directive (now Regulations). To identify cosmetic product types containing chlorhexidine, and to measure the concentration of chlorhexidine in selected products. Between February 2013 and April 2013, we checked for chlorhexidine in cosmetic products in 14 supermarkets, one hairdressing salon and one beauty and retail store in Copenhagen, Denmark by reading the ingredient labels. The chlorhexidine concentration was measured in 10 selected products by high-performance liquid chromatography (HPLC) with an ultraviolet (UV) detector. Chlorhexidine was found in 80 of 2251 checked products (3.6%) in the following categories: hair products (57/760), creams (9/324), face washes (4/24), wet wipes (4/63), skin tonics (3/22), make-up removers (2/25), and mouth washes (1/17). Chlorhexidine concentrations were 0.01-0.15%. We found chlorhexidine in various cosmetic product types, predominantly aimed at females, and in hair products. The measured chlorhexidine concentrations were all within the permitted limit. The relevance for allergic sensitization should be further explored.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Chlorhexidine, ≥99.5%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
USP
Kwas octowy lodowaty, United States Pharmacopeia (USP) Reference Standard
Supelco
Chlorhexidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Chlorhexidine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M