Merck
  • Simultaneous quantification of 25 active constituents in the total flavonoids extract from Herba Desmodii Styracifolii by high-performance liquid chromatography with electrospray ionization tandem mass spectrometry.
Simultaneous quantification of 25 active constituents in the total flavonoids extract from Herba Desmodii Styracifolii by high-performance liquid chromatography with electrospray ionization tandem mass spectrometry.
Journal of separation science (2015-01-27)
Panpan Guo, Wenying Yan, Qingjie Han, Chunying Wang, Zijian Zhang
ABSTRAKT
A sensitive and selective high-performance liquid chromatography coupled with electrospray ionization tandem mass spectrometry method has been developed and validated for the simultaneous determination of 25 active constituents, including 21 flavonoids and four phenolic acids in the total flavonoids extract from Herba Desmodii Styracifolii for the first time. Among the 25 compounds, seven compounds including caffeic acid, acacetin, genistein, genistin, diosmetin, diosmin and hesperidin were identified and quantified for the first time in Herba Desmodii Styracifolii. Chromatographic separation was accomplished on a ZORBAX SB-C18 (250 mm×4.6 mm, 5.0 μm) column using gradient elution of methanol and 0.1‰ acetic acid v/v at a flow rate of 1.0 mL/min. The identification and quantification of the analytes were achieved using negative electrospray ionization mass spectrometry in multiple-reaction monitoring mode. The method was fully validated in terms of limits of detection and quantification, linearity, precision and accuracy. The results indicated that the developed method is simple, rapid, specific and reliable. Furthermore, the developed method was successfully applied to quantify the 25 active components in six batches of total flavonoids extract from Herba Desmodii Styracifolii.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Alkohol odwodniony, United States Pharmacopeia (USP) Reference Standard
Diosmin, European Pharmacopoeia (EP) Reference Standard
Supelco
Genistein, analytical standard
Sigma-Aldrich
Diosmetin
Supelco
Apigenin, analytical standard
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)
Sigma-Aldrich
Acacetin, ≥97.0% (HPLC)
Supelco
Diosmin, analytical standard
Sigma-Aldrich
Naringenin, natural (US), 98%
Sigma-Aldrich
(±)-Naringenin, ≥95%
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Supelco
(±)-Naringenin, analytical standard
Diosmin for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Luteolin, analytical standard
Supelco
Hesperidin, analytical standard
Sigma-Aldrich
Hesperidin, ≥80% (HPLC)
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Supelco
3-glukozyd kwercetyny, analytical standard
Hesperidin, European Pharmacopoeia (EP) Reference Standard
Luteolin, primary reference standard
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Isovitexin, primary reference standard
Izokwercytryna, primary reference standard
Sigma-Aldrich
Isovitexin, ≥98.0% (HPLC)
Hesperidin, primary reference standard