Merck
Cell selective chitosan microparticles as injectable cell carriers for tissue regeneration.
Biomaterials (2015-01-17)
C A Custódio, M T Cerqueira, A P Marques, R L Reis, J F Mano
ABSTRAKT
The detection, isolation and sorting of cells holds an important role in cell therapy and regenerative medicine. Also, injectable systems have been explored for tissue regeneration in vivo, because it allows repairing complex shaped tissue defects through minimally invasive surgical procedures. Here we report the development of chitosan microparticles with a size of 115.8 μm able to capture and expand a specific cell type that can also be regarded as an injectable biomaterial. Monoclonal antibodies against cell surface antigens specific to endothelial cells and stem cells were immobilized on the surface of the microparticles. Experimental results showed that particles bioconjugated with specific antibodies provide suitable surfaces to capture a target cell type and subsequent expansion of the captured cells. Primarily designed for an application in tissue engineering, three main challenges are accomplished with the herein presented microparticles: separation, scale-up expansion of specific cell type and successful use as an injectable system to form small tissue constructs in situ.
MATERIAŁY
Numer produktu
Marka
Opis produktu
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium bicarbonate, ACS reagent, 99.7%, powder, crystals or granules
Sigma-Aldrich
Potassium bicarbonate, ≥99.95% trace metals basis, 99.7-100.5% dry basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Wodorowęglan sodu, −40-+140 mesh, ≥95%
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Ammonium chloride, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
3,3′-Dioctadecyloxacarbocyanine perchlorate
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Potassium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Ammonium chloride, BioUltra, ≥99.5% (AT)