Merck
Phytochemical profiles and antioxidant activities in six species of ramie leaves.
PloS one (2014-09-23)
Yongsheng Chen, Gaoyan Wang, Hong Wang, Chaohua Cheng, Gonggu Zang, Xinbo Guo, Rui Hai Liu
ABSTRAKT
Increased consumption of vegetables or plant food has been associated with decreased risk of developing major chronic diseases, such as cancers, diabetes, cardiovascular diseases, and age-related functional decline. Ramie leaves are rich in phenolics and flavonoids, which have been suggested for human health benefits. Phenolic contents, flavonoid contents, phenolic compounds, and anti-cancer properties in six species of ramie leaves were analyzed by Folin-reagent method, sodium borohydride/chloranil-based assay (SBC), HPLC method and antiproliferation, cytoxicity, respectively. Antioxidant activities were measured through peroxyl radical scavenging capacity (PSC) method, oxygen radical absorbance capacity (ORAC) method, and cellular antioxidant activity (CAA). Research indicated that Boehmeria penduliflora contained the highest total phenolic content (2313.7±27.28 mg GAE/100 g FW), and flavonoid content (1682.4±27.70 mg CAE/100 g FW). Boehmeria tricuspis showed the highest PSC value (9574.8±117.63 µM vit. C equiv./100 g FW), while Boehmeria penduliflora indicated the highest ORAC value (330.44±16.88 µmol Trolox equiv./g FW). The antioxidant activities were correlated with phenolic contents and flavonoid contents. Boehmeria tricuspis had the highest antiproliferative capacity with the lowest EC₅₀ (4.11±0.19 mg/mL). The results for the analyzed ramie for CAA were significantly different from each other (p<0.05), Boehmeria tricuspis had the highest CAA value (133.63±7.10 µmol QE/100 g). Benzoic acid, 4-coumaric acid, caffeic acid, and ferulic acid were the dominant phenolic ingredients in the ramie leaves according to HPLC analysis. Our research is the first report to study the phytochemical profiles and antioxidant activities in different species of ramie leaves for their health benefit.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Standardowa gęstość 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Sodium 2,6-dichloroindophenolate hydrate, ACS reagent
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Deionized
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Standardowa temperatura topnienia waniliny, United States Pharmacopeia (USP) Reference Standard
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Supelco
Vanillin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, PCR Reagent
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline