Merck
Phytochemical composition and antinociceptive activity of Bauhinia glauca subsp. hupehana in rats.
PloS one (2015-02-07)
Jinlong Xu, Qizhi Zhao, Lei Wei, Yu Yang, Rui Xu, Nengjiang Yu, Yimin Zhao
ABSTRAKT
In traditional medicine, Bauhinia glauca subsp. hupehana has long been used as an analgesic agent in China. The aim of this study was to evaluate the antinociceptive activity of the ethanol extract of the aerial parts of B. glauca subsp. hupehana (BHE) in rats and its chemical fingerprint. The antinociceptive activity of BHE was assessed in mice using chemically and heat-induced pain models, such as the acetic acid-induced writhing, hot plate, tail-flick and glutamate tests. Naltrexone hydrochloride, a non-selective opioid receptor antagonist, was utilized to determine the involvement of the opioid system. In addition to this, the involvements of the cGMP and ATP-sensitive K+ channel pathways were also detected using methylene blue and glibenclamide. The oral administration of BHE (at doses of 50, 100 and 200 mg/kg) produced significant and dose-related inhibitions in both the chemically and heat-induced pain models. Interestingly, in the abdominal constriction test, when the dose of BHE was increased to 800 mg/kg (p.o., n = 10), the inhibition rate was 100%. The antinociceptive mechanism may involve the cGMP pathway and ATP sensitive K+ channel pathway. The central antinociceptive effect was not antagonized by naltrexone. One phenolic acid, one lignin and five flavonoids were isolated from BHE. The antinociceptive activity of BHE was most likely due to the presence of the flavonoids. The acute toxicity results showed that BHE was safe at a high dose (2 g/kg, p.o.). The current investigation demonstrates that B. glauca subsp. hupehana is a potential candidate for the development of novel, non-opioid, analgesic phytomedicines.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Naltrexone hydrochloride
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Naltrexone hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology