Merck
Expression and colocalization patterns of calbindin-D28k, calretinin and parvalbumin in the rat hypothalamic arcuate nucleus.
Journal of chemical neuroanatomy (2014-07-12)
Kylie S Foo, Arash Hellysaz, Christian Broberger
ABSTRAKT
Calcium binding proteins (CaBPs) form a diverse group of molecules that function as signal transducers or as intracellular buffers of Ca(2+) concentration. They have been extensively used to histochemically categorize cell types throughout the brain. One region which has not yet been characterized with regard to CaBP expression is the hypothalamic arcuate nucleus, which plays a vital role in neuroendocrine control and the central regulation of energy metabolism. Using in situ hybridization and immunofluorescence, we have investigated the cellular distribution of the three CaBPs, calbindin-D28k (CB), calretinin (CR) and parvalbumin (PV) in the rat arcuate nucleus. Both mRNA and immunoreactivity was detected in the arcuate nucleus for CB - located in the medial aspects - and CR - located ventrolaterally. No PV mRNA was detected in the arcuate nucleus. Immunofluorescence results for PV were ambiguous; while one antibody detected a group of cell somata, a different antibody failed to visualize any arcuate nucleus cell profiles. Using double-labeling, neither of the examined CaBPs were observed in cells immunoreactive for the signaling molecules agouti gene-related protein, tyrosine hydroxylase, neurotensin, growth hormone-releasing hormone, somatostatin, enkephalin, dynorphin or galanin. We did, however, observe CB- and CR-immunoreactivity, in two distinct populations of neurons immunoreactive for the melanocortin peptide α-melanocyte-stimulating hormone. These data identify distinct subpopulations of arcuate neurons defined by their expression of CaBPs and provide further support for differentiation between subpopulations of anorexigenic melanocortin neurons.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
N-Lauroylsarcosine, neat, ≥95%
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Dabco® 33-LV
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Neurotensin, ≥90% (HPLC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
N-Lauroylsarcosine, purum p.a., ≥98.0% (GC)
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)