Merck
A human monoclonal antibody targeting the stem cell factor receptor (c-Kit) blocks tumor cell signaling and inhibits tumor growth.
Cancer biology & therapy (2014-06-13)
Maria B Lebron, Laura Brennan, Christopher B Damoci, Marie C Prewett, Marguerita O'Mahony, Inga J Duignan, Kelly M Credille, James T DeLigio, Marina Starodubtseva, Michael Amatulli, Yiwei Zhang, Kaben D Schwartz, Douglas Burtrum, Paul Balderes, Kris Persaud, David Surguladze, Nick Loizos, Keren Paz, Helen Kotanides
ABSTRAKT
Stem cell factor receptor (c-Kit) exerts multiple biological effects on target cells upon binding its ligand stem cell factor (SCF). Aberrant activation of c-Kit results in dysregulated signaling and is implicated in the pathogenesis of numerous cancers. The development of more specific and effective c-Kit therapies is warranted given its essential role in tumorigenesis. In this study, we describe the biological properties of CK6, a fully human IgG1 monoclonal antibody against the extracellular region of human c-Kit. CK6 specifically binds c-Kit receptor with high affinity (EC 50 = 0.06 nM) and strongly blocks its interaction with SCF (IC 50 = 0.41 nM) in solid phase assays. Flow cytometry shows CK6 binding to c-Kit on the cell surface of human small cell lung carcinoma (SCLC), melanoma, and leukemia tumor cell lines. Furthermore, exposure to CK6 inhibits SCF stimulation of c-Kit tyrosine kinase activity and downstream signaling pathways such as mitogen-activated protein kinase (MAPK) and protein kinase B (AKT), in addition to reducing tumor cell line growth in vitro. CK6 treatment significantly decreases human xenograft tumor growth in NCI-H526 SCLC (T/C% = 57) and Malme-3M melanoma (T/C% = 58) models in vivo. The combination of CK6 with standard of care chemotherapy agents, cisplatin and etoposide for SCLC or dacarbazine for melanoma, more potently reduces tumor growth (SCLC T/C% = 24, melanoma T/C% = 38) compared with CK6 or chemotherapy alone. In summary, our results demonstrate that CK6 is a c-Kit antagonist antibody with tumor growth neutralizing properties and are highly suggestive of potential therapeutic application in treating human malignancies harboring c-Kit receptor.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Dacarbazine, European Pharmacopoeia (EP) Reference Standard
USP
Transplatyna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Dacarbazine, antineoplastic purine analog
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Etoposide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride