Merck
Collagenase digestion down-regulates the density of CD27 on lymphocytes.
Journal of immunological methods (2014-07-30)
Zhangbo Chen, Xiangyu Chen, Yong Xu, Ping Xiong, Min Fang, Zheng Tan, Feili Gong, Fang Zheng
ABSTRAKT
Collagenases are widely used for tissue digestion in experiments due to their potent hydrolysis of connective tissue. CD27, also known as tumor necrosis factor receptor superfamily 7 (TNFRSF7), is limited to be expressed on the cells of the lymphoid lineage. In our preliminary research, we found that CD27 on NK cells was almost disappeared with the digestion of type I collagenase for 90min. This phenomenon suggests that the process of tissue digestion may affect the density of CD27 on cells. In order to verify this, the lungs of mice were digested with types I and IV collagenase or grinded, respectively. The percentage of CD27(+) cells and the density of CD27 on cells were assayed by flow cytometry. The data presented that the percentage of CD27(+) cells and the density of CD27 on lymphocytes gradually decreased with the time of digestion with type I or IV collagenase. We also detected that the density of CD11b on NK cells was not affected by collagenase digestion. Collectively, the findings of the present study suggest that the collagenase digestion has a selective effect on the density of molecules on cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder