Merck
Resveratrol attenuates the inflammatory reaction induced by ischemia/reperfusion in the rat heart.
Molecular medicine reports (2014-04-01)
Xiaoqiang Cong, Ying Li, Na Lu, Yajian Dai, Huijie Zhang, Xin Zhao, Ya Liu
ABSTRAKT
The role of resveratrol (Res) in inflammation induced by ischemia/reperfusion is not well understood. The aim of the present study was to investigate whether Res modulates neutrophil accumulation and tumor necrosis factor-α (TNF-α) induction in an ischemia/reperfusion-injured rat heart model. The rats were randomly exposed to sham surgery, myocardial ischemia/reperfusion (MI/R) alone, MI/R + Res, MI/R + Res + L-NG-nitroarginine methyl ester (L-NAME) and MI/R + Res + methylene blue (MB). The results demonstrated that compared with MI/R, Res reduced the myocardial infarct area, myocardial myeloperoxidase levels, serum creatinine kinase and lactate dehydrogenase levels, and serum and myocardial TNF-α production. All the effects of Res demonstrated were inhibited by L-NAME (a nitric oxide (NO) synthase inhibitor) and MB [a cyclic guanosine monophosphate (cGMP) inhibitor]. Thus, Res produces cardioprotective and anti-inflammatory effects. These effects may be associated with an increase in NO production, the inhibition of neutrophil accumulation, TNF-α induction and cGMP signaling pathways in myocardium subjected to MI/R.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology