Merck
Development and validation of an immunochromatographic assay for rapid detection of fumonisin B1 from cereal samples.
Journal of food science and technology (2014-09-06)
M Venkataramana, K Navya, S Chandranayaka, S R Priyanka, H S Murali, H V Batra
ABSTRAKT
Fumonisins are one of the most agriculturally significant environmental toxins produced by Fusarium and Aspergillus species that grow on agricultural commodities in the field or during storage. Cereals contaminated with fumonisins causes serious loss to agricultural produce leads to health problems in humans and other farm animals. In the present study, polyclonal hyperimmune sera was raised against FB1 in rabbits immunized with FB1-keyhole limpet haemocyanin (KLH). Purified antibodies were used to establish a sensitive gold nanoparticle based immunochromatographic strip (ICG) for detecting FB1 levels in cereal grains. Effective on-site detection of FB1 was achieved by developing a rapid and sensitive pAb based ICG strip. This strip had a detection limit of 5 ng mL(-1) for FB1 in cereal samples and it could be completed within 3 min. Close examination of 150 cereal samples by ICG strip method revealed that 77 were fumonisin-positive. Results obtained by the developed method was further validated with well standardized HPLC method and results of strip method was correlated well with those obtained by HPLC method. In conclusion, the developed method was a better alternative for onsite detection of FB1 in cereal samples intended for human consumption to reduce risk of humans and other farm animals. The high level of FB1 concentrations recorded in present study warrants the need to develop an awareness creation programme to the farmers of India for safe handling of cereal grains at the time of harvesting and storage of grains.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Deoxynivalenol
Sigma-Aldrich
Zearalenone, fungal mycotoxin
Sigma-Aldrich
T-2 Toxin, from Fusarium sp., powder, ≥98% (HPLC)
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium sulfate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Supelco
T2-Toxin solution, 100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Supelco
Fumonisin B1 solution, ~50 μg/mL in acetonitrile: water, analytical standard
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Fumonisin B1 from Fusarium moniliforme, ≥98% (HPLC)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Fumonisin B1, reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland