Merck
Pharmacokinetics and excretion of (14)C-lenvatinib in patients with advanced solid tumors or lymphomas.
Investigational new drugs (2014-11-08)
Anne-Charlotte Dubbelman, Hilde Rosing, Cynthia Nijenhuis, Alwin D R Huitema, Marja Mergui-Roelvink, Anubha Gupta, David Verbel, Gary Thompson, Robert Shumaker, Jan H M Schellens, Jos H Beijnen
ABSTRAKT
Lenvatinib is an orally available multi-targeted tyrosine kinase inhibitor with anti-angiogenic and antitumor activity. To get more insight into the disposition of lenvatinib, a mass balance study was performed in patients with advanced solid tumors. A single oral 24 mg (100 μCi) dose of (14)C-lenvatinib was administered to six patients, followed by collection of blood, plasma, urine and feces for 7 to 10 days. The collected material was analyzed for total radioactivity, unchanged lenvatinib and selected metabolites. The safety and antitumor effect of a daily oral dose of 24 mg non-labeled lenvatinib were assessed in the extension phase of the study. Peak plasma concentrations of lenvatinib and total radioactivity were reached 1.6 and 1.4 h after administration, respectively, and their terminal phase half-lifes were 34.5 and 17.8 h, respectively. Unchanged lenvatinib systemic exposure accounted for 60 % of the total radioactivity in plasma. Peak concentrations of the analyzed metabolite were over 700-fold lower than the peak plasma concentration of lenvatinib. Ten days after the initial dose, the geometric mean (± CV) recovery of administered dose was 89 % ±10 %, with 64 % ±11 % recovered in feces and 25 % ±18 % in urine. Unchanged lenvatinib in urine and feces accounted for 2.5 % ±68 % of the administered dose, indicating a major role of metabolism in the elimination of lenvatinib. In conclusion, lenvatinib is rapidly absorbed and extensively metabolized, with subsequent excretion in urine and, more predominantly, in feces. Additionally, lenvatinib showed acceptable safety and preliminary antitumor activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Warfarin, analytical standard
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Warfarin, PESTANAL®, analytical standard
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
USP
Warfarin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)