Merck
Surface hydroxyl groups direct cellular response on amorphous and anatase TiO2 nanodots.
Colloids and surfaces. B, Biointerfaces (2014-09-13)
Yi Hong, Mengfei Yu, Jun Lin, Kui Cheng, Wenjian Weng, Huiming Wang
ABSTRAKT
In this study, we investigated the differences between amorphous and anatase TiO2 at the biomolecular level which could explain differences in the osteoblast response on these surfaces. The number of surface hydroxyl groups in the TiOHT form on amorphous and anatase TiO2 was found to be the most important factor, resulting in adsorption of bovine serum albumin as a monolayer on amorphous TiO2 nanodots but as a multilayer on anatase TiO2 nanodots. The reason for this is that the presence of more TiOHT groups on amorphous TiO2 nanodots attracts more -NH3+ groups on BSA molecules, causing the conformation of surface-bound BSA molecules to differ from those adsorbed on anatase TiO2 nanodots. Fibronectin which is subsequently adsorbed on anatase TiO2 nanodots then retains a more active conformation for osteoblast adhesion and mineralization.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Cetylpyridinium chloride, European Pharmacopoeia (EP) Reference Standard
USP
Chlorek cetylopirydyniowy, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
4-Methylumbelliferyl phosphate disodium salt, phosphatase substrate
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Titanium(IV) butoxide, purum, ≥97.0% (gravimetric)
Sigma-Aldrich
Titanium(IV) butoxide, reagent grade, 97%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Cetylpyridinium chloride, meets USP testing specifications
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%