Merck
Four types of fatty acids exert differential impact on pancreatic cancer growth.
Cancer letters (2015-02-14)
Ming Yu, Hongyi Liu, Yijie Duan, Dapeng Zhang, Shasha Li, Feng Wang
ABSTRAKT
Increased fatty acids (FAs) regulate pancreatic cancer progression, however, the detailed mechanism is not clear, and different forms of FAs may play diversified roles in pancreatic cancer. To elucidate the underlying mechanism, we compared the effects of four major types of FAs on pancreatic cancer growth both in cell culture and in a mouse model. HPAF pancreatic cancer cells were implanted in nude mice for 14 weeks, and the mice were fed with four different high-fat/high-energy diets (15% fat, 4 kcal/g), an iso-caloric diet (5% fat, 4 kcal/g) and a normal diet (4% fat, 3 kcal/g). The high fat diets were rich in saturated fatty acids (SFAs), monounsaturated fatty acids (MUFAs), and n-6 and n-3 polyunsaturated fatty acids (n6- and n3PUFAs), respectively. While n3PUFA diet decreased tumor viability, the other high fat diets stimulated tumor viability by apparently different mechanisms. For instance, xenografts whose carriers were fed with SFA diet had marked expression of cancer-related proteins and lipid droplets. Although mice that were fed with MUFA- and n6PUFA diets had pancreatic tumors of similar size, liver metastasis occurred more frequently in those with the n6PUFA diet. In experiments in vitro, the HPAF-cell population was increased by SFAs and MUFAs, decreased by n3PUFAs and not changed by n6PUFAs. In conclusion, different fatty acids have different impact on pancreatic cancer cells. The effects of fatty acids on pancreatic cancer cells were consistent in vivo and in vitro except that n6PUFAs only had regulatory effects in vivo.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
(L)-Dehydroascorbic acid
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Dehydro-L-(+)-ascorbic acid dimer, ≥80% (enzymatic)
Sigma-Aldrich
Dodecanoic acid, 98%
Supelco
Dodecanoic acid, analytical standard
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitoleic acid, ≥98.5% (GC), liquid
Sigma-Aldrich
Palmitic acid, ≥99%
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Lauric acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Palmitoleic acid, analytical standard
Supelco
Palmitic acid, analytical standard
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture