Merck
Molecular detection and characterization of BK and JC polyomaviruses in urine samples of renal transplant patients in Southern Brazil.
Journal of medical virology (2015-01-22)
Juliana Comerlato, Fabrício Souza Campos, Martha Trindade Oliveira, Samuel Paulo Cibulski, Lilian Corrêa, Marcos Iuri Roos Kulmann, Thalita Souza Arantes, Lucas Peretti Hentges, Fernando Rosado Spilki, Paulo Michel Roehe, Ana Cláudia Franco
ABSTRAKT
The human polyomaviruses JC (JCPyV) and BK (BKPyV) are widespread in the human population. Following the primary infection, virus reactivation may lead to nephropathy and graft rejection in renal transplant patients. This study was carried out to access the presence of BKPyV and JCPyV DNA in urine samples collected from renal transplant patients (n = 92) and healthy individuals (n = 88) in Porto Alegre, Rio Grande do Sul. The samples were submitted to a nested PCR. A significantly higher frequency (P < 0.001) of BKPyV was found in renal transplant patients (65.2%) in comparison to the control group (32.9%). JCPyV was detected equally in both groups. Phylogenetic analysis of both BKPyV and JCPyV amplicons demonstrates the presence of the BKPyV subtypes I and II, whereas for JCPyV, four different groups are found (1, 2, 3, and 4).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)