Merck
Impaired vitamin K recycling in uremia is rescued by vitamin K supplementation.
Kidney international (2014-01-17)
Nadine Kaesler, Elke Magdeleyns, Marjolein Herfs, Thomas Schettgen, Vincent Brandenburg, Danilo Fliser, Cees Vermeer, Jürgen Floege, Georg Schlieper, Thilo Krüger
ABSTRAKT
In chronic kidney disease, vitamin K-dependent proteins, including the calcification inhibitor matrix Gla protein, are largely uncarboxylated indicating that functional vitamin K deficiency may contribute to uremic vascular calcification. Since the effects of uremia on the vitamin K cycle are unknown, we investigated the influence of uremia and vitamin K supplementation on the activity of the vitamin K cycle and extraosseous calcification. Uremia was induced in rats by an adenine-supplemented diet and vitamin K1 or K2 was administered over 4 and 7 weeks. After 4 weeks of adenine diet, the activity of the vitamin K cycle enzyme γ-carboxylase but not the activities of DT-diaphorase or vitamin K epoxide reductase were reduced. Serum levels of undercarboxylated matrix Gla protein increased, indicating functional vitamin K deficiency. There was no light microscopy-detectable calcification at this stage but chemically determined aortic and renal calcium content was increased. Vitamin K treatment reduced aortic and renal calcium content after 4 weeks. Seven weeks of uremia induced overt calcification in the aorta, heart, and kidneys; however, addition of vitamin K restored intrarenal γ-carboxylase activity and overstimulated it in the liver along with reducing heart and kidney calcification. Thus, uremic vitamin K deficiency may partially result from a reduction of the γ-carboxylase activity which possibly contributes to calcification. Pharmacological vitamin K supplementation restored the vitamin K cycle and slowed development of soft tissue calcification in experimental uremia.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
3,3′-Methylene-bis(4-hydroxycoumarin)