Merck
Silver and gold nanostructures: antifungal property of different shapes of these nanostructures on Candida species.
Medical mycology (2013-08-24)
Ali Jebali, Farzaneh Haji Esmaeil Hajjar, Fereydoun Pourdanesh, Seyedhossein Hekmatimoghaddam, Bahram Kazemi, Alireza Masoudi, Karim Daliri, Najme Sedighi
ABSTRAKT
The shape of nanoparticles is an important determinant of their physical and chemical properties, possibly including the little-explored area of their use as antifungal agents. Therefore, we evaluated the in vitro antifungal activities of three different shapes of silver and gold nanostructures, including nanocubes, nanospheres, and nanowires, on Candida albicans, C. glabrata and C. tropicalis, using the microdilution and disk diffusion methods as per the guidelines of the Clinical and Laboratory Standards Institute. We found that silver and gold nanocubes had higher antifungal properties against the test species than nanospheres and nanowires. While some isolates were resistant to silver and gold nanospheres and nanowires, none of the isolates were resistant to silver and gold nanocubes. The occurrence of resistance is a new finding which should be further explored.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dekstroza, United States Pharmacopeia (USP) Reference Standard