Merck
Dermal xenobiotic metabolism: a comparison between native human skin, four in vitro skin test systems and a liver system.
Skin pharmacology and physiology (2014-06-20)
Christian Wiegand, Nicola J Hewitt, Hans F Merk, Kerstin Reisinger
ABSTRAKT
The xenobiotic metabolism of 4 in vitro human skin test systems (2D and 3D) was compared with that of the native human skin samples from which the skin test systems had been produced. In total 3 skin samples were investigated, each from a different donor to exclude variability due to gender, donor or tissue supplier. In addition, the skin cultures were compared with a surrogate of the liver. Basal and induced phase I and phase II enzymes were analyzed regarding gene/protein expression as well as enzyme activity. The distinctions between the different test systems and the two dermal compartments (epidermis and dermis) were more noticeable than any donor variability. The 3D models of skin and liver mirrored the in vivo situation more realistically than did the monolayer cultures. Phase I metabolism was more pronounced in the hepatic model, whereas phase II metabolism was more prominent in the reconstructed skin. These results show that reconstructed skin models are a valuable tool for organ-specific safety assessment with regard to xenobiotic metabolism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
Tretinoin, European Pharmacopoeia (EP) Reference Standard
USP
Tretynoina, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
3,3′-Methylene-bis(4-hydroxycoumarin)
Sigma-Aldrich
4-Methylumbelliferone, ≥98%
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Supelco
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenine, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hymecromone, European Pharmacopoeia (EP) Reference Standard