Merck
Human RAD23 homolog A is required for the nuclear translocation of apoptosis-inducing factor during induction of cell death.
Biology of the cell (2014-07-25)
Janaki N Sudhakar, Kuan-Chih Chow
ABSTRAKT
During the initiation of cell death, mitochondrial protein, apoptosis-inducing factor (AIF), is transported to the nucleus. The mechanism of AIF nuclear translocation, however, is not clear. After protein synthesis, the AIF is originally targetted to the mitochondria, and the nuclear targetting is a secondary event. Therefore, we hypothesised that the nuclear translocation of AIF could be achieved by a novel pathway. By using yeast two-hybrid assay, we identified the human UV excision repair protein RAD23 homolog A (hHR23A) interacts with AIF and their interaction was confirmed by co-immunoprecipitation and fluorescence resonance energy transfer microscopy. Silencing the RAD23A gene expression inhibits the nuclear transportation of AIF and increases cisplatin resistance. Silencing the karyopherin alpha 2 (KPNA2) gene expression, however, did not affect the nuclear import of AIF. Moreover, 2,4-dinitrophenol inhibits staurosporine-induced nuclear translocation of AIF and increases cisplatin resistance. These results suggest that hHR23A is required for the nuclear translocation of AIF during induction of cell death, and this process is energy dependent, but independent of karyopherins.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
USP
Transplatyna, United States Pharmacopeia (USP) Reference Standard
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Etoposide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Etoposide, A cell-permeable derivative of podophyllotoxin that acts as a topoisomerase II inhibitor (IC50 = 59.2 µM) has major activity against a number of tumors, including germ cell neoplasms, small cell lung cancer, and malignant lymphoma.