Merck
A role for transcription factor glial cell missing 2 in Ca2+ homeostasis in zebrafish, Danio rerio.
Pflugers Archiv : European journal of physiology (2014-06-05)
Yusuke Kumai, Raymond W M Kwong, Steve F Perry
ABSTRAKT
The present study investigated the role of the transcription factor, glial cell missing 2 (gcm2), in Ca(2+) regulation in zebrafish larvae. Translational gene knockdown of gcm2 decreased Ca(2+) uptake and the density of ionocytes expressing the epithelial Ca(2+) channel (ecac), and disrupted the overall Ca(2+) balance. Ca(2+) uptake and the expression of gcm2 messenger RNA (mRNA) were significantly elevated in larvae acclimated to low Ca(2+) water (25 μM); the stimulation of Ca(2+) uptake was not observed in fish experiencing gcm2 knockdown. Acclimation to acidic water (pH 4) significantly reduced whole-body Ca(2+) content owing to reduced Ca(2+) uptake and increased Ca(2+) efflux. However, ecac mRNA levels and the density of ecac-expressing ionocytes were increased in fish acclimated to acidic water, and maximal Ca(2+) uptake capacity (J MAX) was significantly increased when measured in control water (pH ~7.4). Acclimation of larvae to acidic water significantly increased gcm2 mRNA expression, and in gcm2 morphants, no such stimulation in Ca(2+) uptake was observed after their return to control water. Overexpression of gcm2 mRNA resulted in a significant increase in the numbers of ecac-expressing ionocytes and Ca(2+) uptake. These observations reveal a critical role for gcm2 in Ca(2+) homeostasis in zebrafish larvae.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard