Merck
Targeted proteomic analyses of nasal lavage fluid in persulfate-challenged hairdressers with bleaching powder-associated rhinitis.
Journal of proteome research (2014-12-30)
Harriet Mörtstedt, Neserin Ali, Monica Kåredal, Helene Jacobsson, Emelie Rietz, Kerstin Kronholm Diab, Jörn Nielsen, Bo A G Jönsson, Christian H Lindh
ABSTRAKT
Hairdressers have an increased risk for developing airway symptoms, for example, asthma and rhinitis. Persulfates, which are oxidizing agents in bleaching powder, are considered important causal agents for these symptoms. However, the underlying mechanisms are unclear. The aim was therefore to measure proteomic changes in nasal lavage fluid from persulfate-challenged subjects to identify proteins potentially involved in the pathogenesis of bleaching powder-associated rhinitis or candidate effect biomarkers for persulfate. Also, oxidized peptides were measured to evaluate their usefulness as biomarkers for persulfate exposure or effect, for example, oxidative stress. Samples from hairdressers with and without bleaching powder-associated rhinitis were analyzed with liquid chromatography tandem mass spectrometry using selected reaction monitoring to target 246 proteins and five oxidized peptides. Pathway analysis was applied to obtain a functional overview of the proteins. Several proteins involved in biologically meaningful pathways, functions, or disorders, for example, inflammatory responses, oxidative stress, epithelium integrity, and dermatological disorders, changed after the persulfate challenge. A list with nine proteins that appeared to be affected by the persulfate challenge and should be followed up was defined. An albumin peptide containing oxidized tryptophan increased 2 h and 5 h after the challenge but not after 20 min, which indicates that such peptides may be useful as oxidative stress biomarkers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid