Merck
Quantitative proteomic analysis of Huh-7 cells infected with Dengue virus by label-free LC-MS.
Journal of proteomics (2014-07-11)
Victoria Pando-Robles, Juan A Oses-Prieto, Myriam Rodríguez-Gandarilla, Erika Meneses-Romero, Alma L Burlingame, Cesar V F Batista
ABSTRAKT
Dengue is an important and growing public health problem worldwide with an estimated 100million new clinical cases annually. Currently, no licensed drug or vaccine is available. During natural infection in humans, liver cells constitute one of the main targets of dengue virus (DENV) replication. However, a clear understanding of dengue pathogenesis remains elusive. In order to gain a better reading of the cross talk between virus and host cell proteins, we used a proteomics approach to analyze the host response to DENV infection in a hepatic cell line Huh-7. Differences in proteome expression were assayed 24h post-infection using label-free LC-MS. Quantitative analysis revealed 155 differentially expressed proteins, 64 of which were up-regulated and 91 down-regulated. These results reveal an important decrease in the expression of enzymes involved in the glycolytic pathway, citrate cycle, and pyruvate metabolism. This study provides large-scale quantitative information regarding protein expression in the early stages of infection that should be useful for better compression of the pathogenesis of dengue. Dengue infection involves alterations in the homeostasis of the host cell. Defining the interactions between virus and cell proteins should provide a better understanding of how viruses propagate and cause disease. Here, we present for the first time the proteomic analysis of hepatocytes (Huh-7 cells) infected with DENV-2 by label-free LC-MS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O