Merck
Antitumor effects and immune regulation activities of a purified polysaccharide extracted from Juglan regia.
International journal of biological macromolecules (2014-09-30)
Wang Ruijun, Wang Shi, Xia Yijun, Tu Mengwuliji, Zhang Lijuan, Wang Yumin
ABSTRAKT
A water-soluble polysaccharide, named as JRP1, was extracted and fractioned from the epicarp of immature fruit of Juglans mandshurica Maxim. The determination of the monosaccharide composition in JRP1 with gas chromatography (GC) showed that JRP1 was composed of Gal (43.1%), Glu (23.6%), Ara (16.2%), Rha (9.8%) and Fru (7.3%). The results in vitro showed that 25, 50 and 100 μg/mL of JRP1 could present a significant inhibition on the growth of S180 cells, and furthermore, a significant improvement on the proliferation ability of lymphocytes and the phagocytic activity of macrophages. The results in vivo showed that compared with those in the control group, the inhibition rates of different doses of JRP1 on S180 cells in the tumor-bearing mice were 35.3%, 40.6% and 48.1%, respectively, and serum immune cytokine levels such as IL-2, TNF-α and IFN-γ were significantly improved. Our results confirm that JRP1 has the activities of effective antitumor and immunomodulatory function.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Fruktoza, United States Pharmacopeia (USP) Reference Standard
Fructose, European Pharmacopoeia (EP) Reference Standard
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
L-(+)-Arabinose, 99%
Sigma-Aldrich
L-(+)-Arabinose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
L-(+)-Arabinose, 98%
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
USP
Galaktoza, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%