Merck
The combined effect of furosemide and propranolol on GSH homeostasis in ACHN renal cells.
Toxicology mechanisms and methods (2014-05-23)
Fatemeh Zal, Raheleh Taheri, Fatemeh Khademi, Ehsan Keshavarz, Sadegh Rajabi, Zohreh Mostafavi-Pour
ABSTRAKT
Propranolol, a beta-adrenergic blocker, is used in the treatment of a large number of cardiovascular diseases such as hypertension and arrhythmias. Propranolol, in combination with furosemide, is used to treat hypertensive disorders although their side effect profile is not very obvious. In present study, the effects of the drugs furosemide and propranolol were in corporately investigated both on glutathione homeostasis and their antioxidant effect on ACHN cells. The cytoxicities and antioxidant effects of these two clinically important drugs on human kidney cell lines were evaluated using MTT following by the determination of glutathione reductase (GR) and glutathione peroxidase (GPx) activities and measuring the level of reduced glutathione (GSH). Propranolol induced a significant cytotoxic effect at 100 µM, while furosemide was cytotoxic at doses of 250 and 1000 µg/ml. A slight increase in GPx and GR activities and GSH level was observed with propranolol and furosemide treatment alone, while the two drugs together caused a significant increase in GPx and GR activities (35% and 42%, respectively) and GSH content (35%) in ACHN cell lysates (p < 0.05). Our results demonstrate that although high doses of furosemide and propranolol are cytotoxic, co-administration of low doses may improve the antioxidant defense in patients undergoing treatment with these two important drugs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%