Merck
Hormonal and metabolic responses to a resistance exercise protocol in lean children, obese children and lean adults.
Pediatric exercise science (2014-11-06)
Daniela A Rubin, Diobel M Castner, Hoang Pham, Jason Ng, Eric Adams, Daniel A Judelson
ABSTRAKT
During childhood, varying exercise modalities are recommended to stimulate normal growth, development, and health. This project investigated hormonal and metabolic responses triggered by a resistance exercise protocol in lean children (age: 9.3 ± 1.4 y, body fat: 18.3 ± 4.9%), obese children (age: 9.6 ± 1.3 y, body fat: 40.3 ± 5.2%) and lean adults (age: 23.3 ± 2.4 y, body fat: 12.7 ± 2.9%). The protocol consisted of stepping onto a raised platform (height = 20% of stature) while wearing a weighted vest (resistance = 50% of lean body mass). Participants completed 6 sets of 10 repetitions per leg with a 1-min rest period between sets. Blood samples were obtained at rest preexercise, immediately postexercise and 2 times throughout the 1-hr recovery to analyze possible changes in hormones and metabolites. Children-adult differences included a larger exercise-induced norepinephrine increase in adults vs. children and a decrease in glucagon in children but not adults. Similarities between adults and children were observed for GH-IGF-1 axis responses. Metabolically, children presented with lower glycolytic and increased fat metabolism after exercise than adults did. Obesity in childhood negatively influenced GH, insulin, and glucose concentrations. While adults occasionally differed from children, amount of activated lean mass, not maturation, likely drove these dissimilarities.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Gliceryna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Glycerol, FCC, FG