Merck
Determination of acetylsalicylic acid and its major metabolites in bovine urine using ultra performance liquid chromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-02-11)
M L Castillo-García, M P Aguilar-Caballos, A Gómez-Hens
ABSTRAKT
A new method based on ultra high performance liquid chromatography (UPLC) with photometric and fluorometric detection for the determination of acetylsalicylic acid and its main metabolites, namely gentisic, salicylic and salicyluric acids, in bovine urine samples is reported. Photometric detection was used for acetylsalicylic acid determination, whereas the native fluorescence of the metabolites was monitored using fluorometric detection. The separation was performed under isocratic conditions, using acetonitrile-phosphate solution (3.5mM, pH 3.5) (26:74, v/v) as the mobile phase. The retention times of the four compounds were lower than 2min, which are shorter than those achieved using conventional HPLC. Under the optimum separation conditions, the dynamic ranges and detection limits (ngmL(-1)) were: 0.2-2500, 0.09 for gentisic acid; 0.2-2500, 0.08 for salicylic acid and 2.5-15,000, 1.1 for salicyluric acid, using fluorescence detection, and 10-25,000, 2.2 for acetylsalicylic acid, using UV detection. Intra-day and inter-day precision data were assessed at two levels of concentration of each analyte using both detection systems. The selectivity of the method was checked by assaying different drugs of veterinary use showing that most of them did not interfere with the determination of the analytes. The method has been applied to the analysis of bovine urine samples, which only required a simple clean up step of the samples prior to injection in the UPLC system. A recovery study was performed, which provided values in the range of 80-100%. This fact proves the practical usefulness of this method as an ultrafast analytical tool for the therapeutic control of acetylsalicylic acid administration in bovine animals intended for food production.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Supelco
2,5-Dihydroxybenzoic acid, analytical standard
USP
Aspiryna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC