Merck
BCL2 promotor methylation and miR-15a/16-1 upregulation is associated with sanguinarine-induced apoptotic death in rat HSC-T6 cells.
Journal of pharmacological sciences (2015-02-24)
Da-Sheng Zhang, Yuan-Yi Li, Xiao-Jun Chen, Yu-Juan Li, Zhao-Ying Liu, Wen-Jian Xie, Zhi-Liang Sun
ABSTRAKT
Previous studies show that several pathways are involved in sanguinarine-induced apoptotic cell death, including AKT downregulation, inhibition of NF-kB activation, mediation of ROS production, downregulation of anti-apoptosis proteins XIAP and cIAP-1, upregulation of BAX, and downregulation of BCL2. In this study, we found out that the quenching of ROS generation by N-acetyl-l-cysteine (NAC), a scavenger of ROS, reversed sanguinarine-induced apoptosis effects, also we found out that sanguinarine-induced rat hepatic stellate T6 cells (HSC-T6 cells) apoptosis was correlated with the generation of increased ROS, which was followed by the activation of caspase-8 (-3, -6, and -9), and the decreasing in the miltochondrial membrane potential (MMP) and the down-regulation of anti-apoptotic protein Bcl-2. It is not clear whether BCL2's downregulation relates to its promoter methylation and miR-15a/16-1 expression which can bind to BCL2 3'-UTR (un-translation reagon). We showed that sanguinarine-induced down regulation of BCL2 was associated with the increased methylation rate of BCL2 promotor district and the increased expression of miR-15a/16-1. HSC-T6 cells treatment with 5-Aza-2'-deoxycytidine (5'-Aza-CdR) impeded sanguinarine-induced BCL2 promotor district methylation and recovered BCL2's expression. Over expression of BCL2 using pEGFP-N1 vector decreased sanguinarine-induced HSC-T6 cells apoptotic death significantly but not completely. These observations clearly showed that BCL2 down regulation was associated with its promoter methylation and miR-15a/16-1 upregulation in sanguinarine-induced Rat HSC-T6 cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
3-Methyl-1-butanol, ACS reagent, ≥98.5%
Sigma-Aldrich
3-Methyl-1-butanol, reagent grade, 98%
Supelco
3-Methyl-1-butanol, analytical standard
Supelco
3-Methylbutanol, analytical standard
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
3-Methylbutanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG
Sigma-Aldrich
Isoamyl alcohol, natural, ≥98%, FG