Merck
Synthesis and application of hybrid polymer composites based on silver nanoparticles as corrosion protection for line pipe steel.
Molecules (Basel, Switzerland) (2014-05-21)
Ayman M Atta, Gamal A El-Mahdy, Hamad A Al-Lohedan, Abdurrahman O Ezzat
ABSTRAKT
A facile method was developed to synthesize in high yield dispersed silver nanoparticles (AgNPs) with small particle sizes of less than 10 nm. Silver nitrate was reduced to silver nanoparticles by p-chloroaniline in the presence of polyoxyethylene maleate 4-nonyl-2-propylene-phenol (NMA) as a stabilizer. The produced AgNPs were used to prepare hybrid polymer based on N-isopropylacrylamide (NIPAm), 2-acrylamido-2-methylpropane sulfonic acid (AMPS), N,N-methylenebisacrylamide (MBA) and potassium persulfate (KPS) using a semi-batch solution polymerization method. The prepared AgNPs and hybrid polymer were characterized by Fourier transform infrared (FTIR) spectroscopy, X-ray diffraction (XRD) patterns and transmission electron microscopy (TEM). The corrosion inhibition activity of the AgNPs and hybrid polymer towards steel corrosion in the presence of hydrochloric acid has been investigated by polarization and electrochemical impedance spectroscopy (EIS) methods. Polarization measurements indicate that the AgNPs and hybrid polymer acts as a mixed type-inhibitor and the inhibition efficiency increases with inhibitor concentration. The results of potentiodynamic polarization and EIS measurements clearly showed that the inhibition mechanism involves blocking of the steel surface by inhibitor molecules via adsorption.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
N-Isopropylacrylamide, ≥99%
USP
Związek G związany z walacyklowirem, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Maleic anhydride, puriss., ≥99.0% (NT)
Sigma-Aldrich
Maleic anhydride, 99%
Supelco
4-Chloroaniline, PESTANAL®, analytical standard
Sigma-Aldrich
4-Chloroaniline, purified by sublimation, ≥99%
Sigma-Aldrich
4-Chloroaniline, 98%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
2-Acrylamido-2-methyl-1-propanesulfonic acid, 99%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer