Merck
Hc-fau, a novel gene regulating diapause in the nematode parasite Haemonchus contortus.
International journal for parasitology (2014-07-25)
Baolong Yan, Xiaolu Guo, Qianjin Zhou, Yi Yang, Xueqiu Chen, Weiwei Sun, Aifang Du
ABSTRAKT
Diapause induced in the early fourth stage of Haemonchus contortus is a strategy to adapt this nematode to hostile environmental conditions. In this study, we identified a new gene, Hc-fau, a homologue of human fau and Caenorhabditis elegans Ce-rps30. Hc-fau encodes two proteins through alternative RNA splicing, Hc-FAUA and Hc-FAUB, consisting of 130 and 107 amino acids, respectively. Hc-FAU possesses a diverged ubiquitin-like (UBiL) protein domain and a conserved ribosome protein S30 domain. The protein is ubiquitously expressed, except in the gonad. However Hc-fau transcripts decrease significantly in diapausing L4s of H. contortus. In C. elegans, knockdown of Ce-rps30 confers an extended lifespan, increased lipid storage in the intestine and shortened body length. These morphological characteristics are comparable with dauer larvae of C. elegans, in which the gonad is condensed considerably. In contrast, a shortened lifespan is observed in C. elegans over-expressing Hc-faua, and especially Hc-faub, with hatching failure detected. The genes of insulin/IGF-1 signalling (IIS), TGF-β, cGMP, dafachronic acid (DA), apoptosis (AP) and fatty acids (FA) metabolism are all down-regulated in Ce-rps30RNAi (RNA interference) worms, except for akt-1 and daf-16. However, daf-16 up-regulation is inconsistent with its target gene down-regulation and the result from a heat stress assay in these worms. Daf-16 RNAi conducted in Ce-rps30 (tm6034/nt1) mutants failed to rescue the worms. The S30 domain stays in the nucleus, while UBiL accumulates in the cytoplasm. Compared with Hc-FAUA, results of UBiL domain and S30 domain over-expression indicate synergism between UBiL and S30 in regulating lifespan and reproduction. These results suggest the potential functions of Hc-fau in regulating larval diapause in H.contortus.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG