Merck
Long-lasting oscillations in the electro-oxidation of formic acid on PtSn intermetallic surfaces.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-05-03)
Nickson Perini, Bruno C Batista, Antonio C D Angelo, Irving R Epstein, Hamilton Varela
ABSTRAKT
Even when in contact with virtually infinite reservoirs, natural and manmade oscillators typically drift in phase space on a time-scale considerably slower than that of the intrinsic oscillator. A ubiquitous example is the inexorable aging process experienced by all living systems. Typical electrocatalytic reactions under oscillatory conditions oscillate for only a few dozen stable cycles due to slow surface poisoning that ultimately results in destruction of the limit cycle. We report the observation of unprecedented long-lasting temporal oscillations in the electro-oxidation of formic acid on an ordered intermetallic PtSn phase. The introduction of Sn substantially increases the catalytic activity and retards the irreversible surface oxidation, which results in the stabilization of more than 2200 oscillatory cycles in about 40 h; a 30-40-fold stabilization with respect to the behavior of pure Pt surfaces. The dynamics were modeled and numerical simulations point to the surface processes underlying the high stability.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%