Merck
Transferrin-mediated fullerenes nanoparticles as Fe(2+)-dependent drug vehicles for synergistic anti-tumor efficacy.
Biomaterials (2014-12-03)
Huijuan Zhang, Lin Hou, Xiaojing Jiao, Yandan Ji, Xiali Zhu, Zhenzhong Zhang
ABSTRAKT
Artesunate (AS) is an iron-dependent drug, which has been used extensively as anti-malarial drugs worldwide with no obvious side effects. Recently, studies have shown that AS also possess profound cytotoxicity against tumor cells. However, simultaneous delivery of hydrophobic AS and Fe(2+) into tumor cells remains a major challenge. Herein, we report a new kind of active-targeting preparations which could not only specially target to tumor cells but also synchronously transfer AS and irons into tumor tissue. In this study, hyaluronic acid (HA) was grafted onto fullerene to get a water-soluble biomaterial (HA-C60) with excellent biocompatibility, and then combined with transferrin (Tf) to obtain a multi-functional drug delivery system (HA-C60-Tf) with significant tumor-targeting efficacy and powerful photodynamic therapy capacity. Finally, AS was adsorbed on HA-C60-Tf with a high loading efficacy of 162.4% (weight ratio of AS: HA-C60-Tf). Compared with free AS, remarkably enhanced antitumor efficacy of AS-loaded HA-C60-Tf nanoparticles was realized both in a cultured MCF-7 cells in vitro and in a tumor-bearing murine model in vivo, due to increased intracellular accumulation of AS in tumor and activated mechanism by co-delivery of Tf and AS analogs. Furthermore, with laser irradiation in vivo, the relative tumor volume (V/V0) of HA-C60-Tf/AS declined by half, from 1.72 ± 0.12 to 0.84 ± 0.07, suggesting a new way with multi-mechanism for tumor treatment was developed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous