Merck
Orexin-1 receptor signaling increases motivation for cocaine-associated cues.
The European journal of neuroscience (2015-03-11)
Brandon S Bentzley, Gary Aston-Jones
ABSTRAKT
The orexin/hypocretin system is involved in multiple cocaine addiction processes that involve drug-associated environmental cues, including cue-induced reinstatement of extinguished cocaine seeking and expression of conditioned place preference. However, the orexin system does not play a role in several behaviors that are less cue-dependent, such as cocaine-primed reinstatement of extinguished cocaine seeking and low-effort cocaine self-administration. We hypothesized that cocaine-associated cues, but not cocaine alone, engage signaling at orexin-1 receptors (OX1Rs), and this cue-engaged OX1R signaling increases motivation for cocaine. Motivation for cocaine was measured in Sprague-Dawley rats with behavioral-economic demand curve analysis after pretreatment with the OX1R antagonist SB-334867 (SB) or vehicle with and without light + tone cues. Demand for cocaine was higher when cocaine-associated cues were present, and SB only reduced cocaine demand in the presence of these cues. We then investigated whether cocaine demand was linked to the cued reinstatement of cocaine seeking, as both procedures are partially driven by cocaine-associated cues in an orexin-dependent manner. SB blocked cue-induced reinstatement behavior, and baseline demand predicted SB efficacy with the largest effect in high-demand animals, i.e. animals with the greatest cue-dependent behavior. We conclude that OX1R signaling increases the reinforcing efficacy of cocaine-associated cues but not that of cocaine alone. This supports our view that orexin plays a prominent role in the ability of conditioned cues to activate motivational responses.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma