Merck
Similarity Evaluation of Different Origins and Species of Dendrobiums by GC-MS and FTIR Analysis of Polysaccharides.
International journal of analytical chemistry (2015-11-06)
Nai-Dong Chen, Nai-Fu Chen, Jun Li, Cai-Yun Cao, Jin-Mei Wang, He-Ping Huang
ABSTRAKT
GC-MS method combined with FTIR techniques by the analysis of polysaccharide was applied to evaluate the similarity between wild (W) and tissue-cultured (TC) Dendrobium huoshanense (DHS), Dendrobium officinale (DO), and Dendrobium moniliforme (DM) as well as 3 wild Dendrobium spp.: Dendrobium henanense (DHN), Dendrobium loddigesii (DL), and Dendrobium crepidatum (DC). Eight monosaccharides involving xylose, arabinose, rhamnose, glucose, mannose, fructose, galactose, and galacturonic acid were identified in the polysaccharide from each Dendrobium sample while the contents of the monosugars varied remarkably across origins and species. Further similarity evaluation based on GC-MS data showed that the r cor values of different origins of DHS, DO, and DM were 0.831, 0.865, and 0.884, respectively, while the r cor values ranged from 0.475 to 0.837 across species. FTIR files of the polysaccharides revealed that the similarity coefficients between W and TC-DHS, DO, and DM were 88.7%, 86.8%, and 88.5%, respectively, in contrast to the similarity coefficients varying from 57.4% to 82.6% across species. These results suggested that the structures of polysaccharides between different origins of the investigated Dendrobiums might be higher than what we had supposed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis