Merck
Zeolitic Core@Shell Adsorbents for the Selective Removal of Free Glycerol from Crude Biodiesel.
ChemSusChem (2015-06-11)
Nima Masoumifard, Pablo M Arnal, Serge Kaliaguine, Freddy Kleitz
ABSTRAKT
Selective adsorption of free glycerol from crude biodiesel was investigated by using mesoporous silica spheres coated with a thin shell of microporous silicalite-1. A polycrystalline silicalite-1 shell was formed upon first covering the external surfaces of various core templates with discrete silicalite-1 nanocrystals, and this was followed by short hydrothermal treatment to ensure shell uniformity. Batch glycerol adsorption experiments were conducted to evaluate the ability of the sorbents to remove free glycerol selectively from crude biodiesel mixtures at various temperatures, also in comparison to that of conventional sorbents, for example, bare mesoporous silica gel spheres and zeolites. The silicalite-1 shell provided a microporous membrane that hindered the diffusion of fatty acid methyl esters into the mesopores of the composite sorbent, whereas the large pore volume of the mesoporous core enabled multilayer glycerol adsorption; this ultimately substantially enhanced the performance in terms of purification yield and adsorption capacity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
Hexadecylamine, 98%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Hexadecylamine, technical grade, 90%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis