Merck
Effect of variation in ovine WFIKKN2 on growth traits appears to be gender-dependent.
Scientific reports (2015-07-23)
Jiqing Wang, Huitong Zhou, Qian Fang, Xiu Liu, Yuzhu Luo, Jon G H Hickford
ABSTRAKT
WFIKKN2 may play a role in the regulation of muscle growth and development, but to date there have been no reports on the effect of variation in WFIKKN2 on growth and carcass traits in livestock. In this study, the effect of variation in ovine WFIKKN2 was investigated in 800 New Zealand Romney lambs (395 male and 405 female), with five previously described variants (A to E) being identified. Variation in ovine WFIKKN2 was not found to affect various growth traits in the female lambs, but the presence of variant B was associated (P < 0.05) with decreased birth weight, tailing weight, weaning weight and pre-weaning growth rate; and increased post-weaning growth rate in male lambs. In male lambs, the presence of variant B was associated (P < 0.05) with an increased shoulder yield and proportion shoulder yield. No associations with growth or carcass traits were detected for the presence (or absence) of the other variants. These results suggest that variation in ovine WFIKKN2 may have a differential effect on growth in male and female lambs, and hence that the gene may be expressed in, or act in, a gender-specific fashion.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.