Merck
Microtubule-bundling activity of the centrosomal protein, Cep169, and its binding to microtubules.
Biochemical and biophysical research communications (2015-10-21)
Yusuke Mori, Yuki Taniyama, Sayori Tanaka, Hiroki Fukuchi, Yasuhiko Terada
ABSTRAKT
CDK5RAP2 is a centrosomal protein that regulates the recruitment of a γ-tubulin ring complex (γ-TuRC) onto centrosomes and microtubules (MTs) dynamics as a member of MT plus-end-tracking proteins (+TIPs). In our previous report, we found mammalian Cep169 as a CDK5RAP2 binding partner, and Cep169 accumulates at the distal ends of MTs and centrosomes, and coincides with CDK5RAP2. Depletion of Cep169 induces MT depolymerization, indicating that Cep169 targets MT tips and regulates stability and dynamics of MTs. However, how Cep169 contributes to the stabilization of MT remains unclear. Here we show that Cep169 is able to stabilize MTs and induces formation of long MT bundles with intense acetylation of MTs with CDK5RAP2, when expressed at higher levels in U2OS cells. In addition, we demonstrated that Cep169 forms homodimers through its N-terminal domain and directly interacts with MTs through its C-terminal domain. Interestingly, Cep169 mutants, which lack each domains, completely abolished the activity, respectively. Therefore, Cep169 bundles MTs and induces solid structure of MTs by crosslinking each adjacent MTs as a homodimer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)