Merck
Ex Vivo Expansion and Differentiation of Human and Mouse Fetal Pancreatic Progenitors Are Modulated by Epidermal Growth Factor.
Stem cells and development (2015-05-01)
Paola Bonfanti, Estelle Nobecourt, Masaya Oshima, Olivier Albagli-Curiel, Veerle Laurysens, Geert Stangé, Mozhdeh Sojoodi, Yves Heremans, Harry Heimberg, Raphael Scharfmann
ABSTRAKT
A comparative analysis of mouse and human pancreatic development may reveal common mechanisms that control key steps as organ morphogenesis and cell proliferation and differentiation. More specifically, understanding beta cell development remains an issue, despite recent progress related to their generation from human embryonic and induced pluripotent stem cells. In this study, we use an integrated approach, including prospective isolation, organ culture, and characterization of intermediate stages, and report that cells from human and mouse fetal pancreas can be expanded in the long term and give rise to hollow duct-like structures in 3D cultures. The expanded cells express a combination of markers (E-cadherin, PDX1, NKX6-1, SOX9, and HNF1β) that reveals pancreatic progenitor identity. Proliferation of embryonic progenitors was stimulated by the Wnt agonist R-spondin1 (RSPO1), FGF10, and EGF. This combination of growth factors allowed maintaining human fetal pancreatic progenitors in culture for many passages, a finding not reported previously. Importantly, in the absence of EGF, proliferation was reduced, while endocrine differentiation was significantly enhanced. We conclude that modulation of EGF signaling affects in vitro expansion and differentiation of progenitors from embryonic pancreas of both mice and man.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Somatostatin, ≥97% (HPLC), powder
Sigma-Aldrich
Glucagon, ≥95% (HPLC), powder, synthetic
Sigma-Aldrich
Somatostatin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Glucagon, synthetic, powder, suitable for cell culture
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)