Merck
Histone deacetylase inhibitor-polymer conjugate nanoparticles for acid-responsive drug delivery.
European journal of medicinal chemistry (2015-04-02)
Iza Denis, Fatima el Bahhaj, Floraine Collette, Régis Delatouche, Fabien Gueugnon, Daniel Pouliquen, Loic Pichavant, Valérie Héroguez, Marc Grégoire, Philippe Bertrand, Christophe Blanquart
ABSTRAKT
We report the synthesis of acid-responsive polymeric nanoparticles (NPs) consisting of a polymer-histone deacetylase inhibitor conjugate. An innovative aspect of this drug delivery particle lies in the NP conjugation of a histone deacetylase (HDAC) inhibitor, CI-994 (Tacedinaline), introduced with a clickable acid-responsive prodrug during monomer synthesis, prior to polymerization. Another novelty lies in the selected norbornene (NB)-polyethylene oxide (PEO) macromonomer allowing standardization of the polymerization process by Ring-Opening Metathesis Polymerization (ROMP) and functionalization through azide-alkyne click chemistry. Herein we demonstrate that the synthesized polymer gave 300 nm core-shell spherical nanoparticles with low dispersity (0.04), high water dispersability thanks to the PEO shell and well controlled HDAC inhibitor prodrug loading. Bioluminescence Resonance Energy Transfer (BRET) assay in living cells and viability experiments demonstrated efficient cellular internalization without additional chemistry, drug release inside cells with restoration of the HDAC inhibition and induction of apoptosis. Such NPs should minimize drug release in vivo during blood circulation and trigger intracellular delivery after endocytosis, holding promises for improved efficacy of this class of epigenetic inhibitors. This standardized synthesis paves the way for multifunctional nanoparticles synthesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Acetone-d6, 99.9 atom % D
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Acetone-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Acetone-d6, "100%", 99.95 atom % D
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Acetone-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Acetone-d6, 99.9 atom % D
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis