Merck
  • Design, synthesis and structure-activity relationship of phthalimides endowed with dual antiproliferative and immunomodulatory activities.
Design, synthesis and structure-activity relationship of phthalimides endowed with dual antiproliferative and immunomodulatory activities.
European journal of medicinal chemistry (2015-05-06)
Marcos Veríssimo de Oliveira Cardoso, Diogo Rodrigo Magalhães Moreira, Gevanio Bezerra Oliveira Filho, Suellen Melo Tiburcio Cavalcanti, Lucas Cunha Duarte Coelho, José Wanderlan Pontes Espíndola, Laura Rubio Gonzalez, Marcelo Montenegro Rabello, Marcelo Zaldini Hernandes, Paulo Michel Pinheiro Ferreira, Cláudia Pessoa, Carlos Alberto de Simone, Elisalva Teixeira Guimarães, Milena Botelho Pereira Soares, Ana Cristina Lima Leite
ABSTRAKT
The present work reports the synthesis and evaluation of the antitumour and immunomodulatory properties of new phthalimides derivatives designed to explore molecular hybridization and bioisosterism approaches between thalidomide, thiosemicarbazone, thiazolidinone and thiazole series. Twenty-seven new molecules were assessed for their immunosuppressive effect toward TNFα, IFNγ, IL-2 and IL-6 production and antiproliferative activity. The best activity profile was observed for the (6a-f) series, which presents phthalyl and thiazolidinone groups.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)