Merck
Bioactive silica nanoparticles reverse age-associated bone loss in mice.
Nanomedicine : nanotechnology, biology, and medicine (2015-02-15)
M Neale Weitzmann, Shin-Woo Ha, Tatyana Vikulina, Susanne Roser-Page, Jin-Kyu Lee, George R Beck
ABSTRAKT
We recently reported that in vitro, engineered 50nm spherical silica nanoparticles promote the differentiation and activity of bone building osteoblasts but suppress bone-resorbing osteoclasts. Furthermore, these nanoparticles promote bone accretion in young mice in vivo. We have now investigated the capacity of these nanoparticles to reverse bone loss in aged mice, a model of human senile osteoporosis. Aged mice received nanoparticles weekly and bone mineral density (BMD), bone structure, and bone turnover were quantified. Our data revealed a significant increase in BMD, bone volume, and biochemical markers of bone formation. Biochemical and histological examinations failed to identify any abnormalities caused by nanoparticle administration. Our studies demonstrate that silica nanoparticles effectively blunt and reverse age-associated bone loss in mice by a mechanism involving promotion of bone formation. The data suggest that osteogenic silica nanoparticles may be a safe and effective therapeutic for counteracting age-associated bone loss. Osteoporosis poses a significant problem in the society. Based on their previous in-vitro findings, the authors' group investigated the effects of spherical silica nanoparticles in reversing bone loss in a mouse model of osteoporosis. The results showed that intra-peritoneal injections of silica nanoparticles could increase bone mineral density, with little observed toxic side effects. This novel method may prove important in future therapy for combating osteoporosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Creatinine, anhydrous, ≥98%