Merck
Purification, characterization, and biological activities of broccolini lectin.
Biotechnology progress (2015-03-05)
Pingping Xu, Ting Zhang, Xiaolei Guo, Chungwah Ma, Xuewu Zhang
ABSTRAKT
Plant lectins have displayed a variety of biological activities. In this study, for the first time, a 27 kDa arabinose- and mannose-specific lectin from Broccolini (Brassica oleracea Italica × Alboglabra), named as BL (Broccolini lectin), was purified by an activity-driven protocol. Mass spectrometry analysis and database search indicated that no matches with any plant lectin were found, but BL contained some peptide fragments (QQQGQQGQQLQQVISR, QQGQQQGQQGQQLQQVISR and VCNIPQVSVCPF QK). BL exhibited hemagglutinating activity against chicken erythrocytes at 4 µg/mL. BL retained full hemagglutinating activity at pH 7-8 and temperature 30-40°C, and had an optimal activity in Ca(2+) solution. Bioactivity assay revealed that BL exhibited dose-dependent inhibition activity on 5 bacterial species with IC50 values of 143.95-486.33 μg/mL, and on 3 cancer cells with IC50 values of 178.82-350.93 μg/mL. Notably, 5-fold reduction in IC50 values was observed on normal L-O2 vs cancerous HepG-2 cells (924.35 vs. 178.82 μg/mL). This suggests that BL should be promising in food and medicine.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)