Merck
Occurrence and distribution of antibiotics in urban soil in Beijing and Shanghai, China.
Environmental science and pollution research international (2015-03-26)
Lihong Gao, Yali Shi, Wenhui Li, Jiemin Liu, Yaqi Cai
ABSTRAKT
The recycling of reclaimed wastewater for irrigation and road cleaning is an important strategy to minimize water scarcity in megacities. However, little is known regarding the potential accumulation of antibiotics contained in reclaimed wastewater in urban soil. We investigated the occurrence and distribution of eight quinolones (QNs), nine sulfonamides (SAs), and five macrolides (MLs) antibiotics in urban surface soil in Beijing and Shanghai, China. QNs, especially norfloxacin (NOR), ofloxacin (OFL), and ciprofloxacin (CIP) were the predominant antibiotics in urban surface soil, and NOR revealed the highest average concentration of 94.6 μg kg(-1). The antibiotic concentrations in urban soil in our study were higher than those detected in agricultural soils after long-term wastewater irrigation and manure fertilization. The concentrations of antibiotics in Shanghai urban soil showed a significant negative correlation with soil pH and a positive correlation with total organic carbon (TOC), reflecting the effect of speciation and soil organic matter content on sorption and retention. In addition, antibiotic concentrations in the urban soil were positively correlated with heavy metal contents, likely due to their coexistence in reclaimed wastewater and the promoting effect of metals on the sorption of antibiotics. In several soil samples, NOR, OFL, CIP, enrofloxacin (ENR), and fleroxacin (FLE) showed higher concentrations than the trigger value of 100 μg kg(-1) in soil, indicating a potential risk for the environment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Sigma-Aldrich
Sulfamonomethoxine
Sigma-Aldrich
Josamycin, ≥90% (HPLC)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Lomefloxacin hydrochloride
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG