Merck
IPP51, a chalcone acting as a microtubule inhibitor with in vivo antitumor activity against bladder carcinoma.
Oncotarget (2015-06-04)
Véronique Martel-Frachet, Michelle Keramidas, Alessandra Nurisso, Salvatore DeBonis, Claire Rome, Jean-Luc Coll, Ahcène Boumendjel, Dimitrios A Skoufias, Xavier Ronot
ABSTRAKT
We previously identified 1-(2,4-dimethoxyphenyl)-3-(1-methylindolyl) propenone (IPP51), a new chalcone derivative that is capable of inducing prometaphase arrest and subsequent apoptosis of bladder cancer cells. Here, we demonstrate that IPP51 selectively inhibits proliferation of tumor-derived cells versus normal non-tumor cells. IPP51 interfered with spindle formation and mitotic chromosome alignment. Accumulation of cyclin B1 and mitotic checkpoint proteins Bub1 and BubR1 on chromosomes in IPP51 treated cells indicated the activation of spindle-assembly checkpoint, which is consistent with the mitotic arrest. The antimitotic actions of other chalcones are often associated with microtubule disruption. Indeed, IPP51 inhibited tubulin polymerization in an in vitro assay with purified tubulin. In cells, IPP51 induced an increase in soluble tubulin. Furthermore, IPP51 inhibited in vitro capillary-like tube formation by endothelial cells, indicating that it has anti-angiogenic activity. Molecular docking showed that the indol group of IPP51 can be accommodated in the colchicine binding site of tubulin. This characteristic was confirmed by an in vitro competition assay demonstrating that IPP51 can compete for colchicine binding to soluble tubulin. Finally, in a human bladder xenograft mouse model, IPP51 inhibited tumor growth without signs of toxicity. Altogether, these findings suggest that IPP51 is an attractive new microtubule-targeting agent with potential chemotherapeutic value.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
Sigma-Aldrich
PIPES, ≥99% (titration)
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
PIPES, BioXtra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
PIPES, anhydrous, free-flowing, Redi-Dri, ≥99%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
D-Serine, ≥98% (TLC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%